1-(4-(Trifluoromethyl)phenyl)-1H-pyrazol-3-amine 97%

Supplier: Ambeed
Warning

A1376698-100MG A1376698-1G
76682-462EA 90.18 USD
76682-462 77597-068
1-(4-(Trifluoromethyl)phenyl)-1H-pyrazol-3-amine 97%
1-(4-(Trifluoromethyl)phenyl)-1H-pyrazol-3-amine

Precaución: for Research Use Only

Formula: C₁₀H₈F₃N₃
MW: 227.1858 g/mol
MDL Number: MFCD16835995
CAS Number: 1207891-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR