N,N-Dibenzyl-3-bromoaniline 97%

Supplier: Ambeed
Warning

A1374826-1G A1374826-250MG
76682-454EA 80 USD
76682-454 76682-456
N,N-Dibenzyl-3-bromoaniline 97%
N,N-Dibenzyl-3-bromoaniline

Precaución: for Research Use Only

Formula: C₂₀H₁₈BrN
MDL Number: MFCD21090540
CAS Number: 148491-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR