6-Amino-2-bromonicotinonitrile 97%

Supplier: Ambeed
Danger

A1372234-100MG A1372234-250MG
76682-432EA 233 USD
76682-432 76682-434
6-Amino-2-bromonicotinonitrile 97%
6-Amino-2-bromonicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₄BrN₃
MW: 198.0201 g/mol
MDL Number: MFCD28715183
CAS Number: 1805270-30-7
UN: 3439
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR