(2S,4R)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-(tert-Butoxycarbonyl)-trans-4-fluoro-L-proline, (2S,4R)-1-Boc-4-fluoro-2-pyrrolidinecarboxylic Acid, (2S,4R)-1-(tert-Butoxycarbonyl)-4-fluoropyrrolidine-2-carboxylic acid, N-Boc-trans-4-fluoro-L-proline
A1371754-100G
A1371754-10G
A1371754-1G
A1371754-25G
A1371754-500G
A1371754-5G
76682-414EA
287
USD
76682-414
76682-416
76682-418
76682-420
76682-422
76682-424
(2S,4R)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid 97%
(2S,4R)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₆FNO₄ |
MDL Number:
MFCD06796085 CAS Number: 203866-14-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...