(S)-2-Benzyl-2,3-dihydrobenzo[d]imidazo[2,1-b]thiazole ≥97%, ee 99%

Supplier: Ambeed
Warning

A1371068-1G A1371068-250MG
76682-400EA 631.24 USD
76682-400 76682-402
(S)-2-Benzyl-2,3-dihydrobenzo[d]imidazo[2,1-b]thiazole ≥97%, ee 99%
(S)-2-Benzyl-2,3-dihydrobenzo[d]imidazo[2,1-b]thiazole

Precaución: for Research Use Only

Formula: C₁₆H₁₄N₂S
MW: 266.3608 g/mol
CAS Number: 1239015-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR