8,8-Difluoro-6-azaspiro[3.4]octane hydrochloride 97%

Supplier: Ambeed
Warning

A1367912-1G A1367912-250MG
76682-394EA 1185.03 USD
76682-394 76682-396
8,8-Difluoro-6-azaspiro[3.4]octane hydrochloride 97%
8,8-Difluoro-6-azaspiro[3.4]octane hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₂ClF₂N
MW: 183.6267 g/mol
MDL Number: MFCD33022454
CAS Number: 2682112-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR