Rhodium, tetrakis[μ-[(αS)-α-(1,1-dimethylethyl)-1,3-dioxo-1H-benz[de]isoquinoline-2(3H)-acetato-κO2:κO2′]]di-, (Rh-Rh)-EA adduct 97%

Supplier: Ambeed
Warning

A1366476-250MG A1366476-50MG A1366476-100MG A1366476-1G
76682-390EA 670 USD
76682-390 76682-392 77867-618 77867-619
Rhodium, tetrakis[μ-[(αS)-α-(1,1-dimethylethyl)-1,3-dioxo-1H-benz[de]isoquinoline-2(3H)-acetato-κO2:κO2′]]di-, (Rh-Rh)-EA adduct 97%
Rhodium, tetrakis[μ-[(αS)-α-(1,1-dimethylethyl)-1,3-dioxo-1H-benz[de]isoquinoline-2(3H)-acetato-κO2:κO2′]]di-, (Rh-Rh)-EA adduct

Precaución: for Research Use Only

CAS Number: 1227093-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR