2-Ethylhexyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate 98%

Supplier: Ambeed
Warning

A1364274-1G A1364274-250MG A1364274-100MG A1364274-25G A1364274-5G
76682-388EA 312 USD
76682-388 77566-710 77592-586 77596-904 77596-906
2-Ethylhexyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate 98%
2-Ethylhexyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate

Precaución: For research use only.

Formula: C₂₅H₄₂O₃
CAS Number: 144429-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR