1-(4-Chloro-3-fluoropyridin-2-yl)ethanone 97%
Supplier: Ambeed
|
Warning
|
A1363620-1G
A1363620-250MG
76682-380EA
176
USD
76682-380
76682-382
1-(4-Chloro-3-fluoropyridin-2-yl)ethanone 97%
1-(4-Chloro-3-fluoropyridin-2-yl)ethanone
Precaución: for Research Use Only
Formula:
C₇H₅ClFNO |
MDL Number:
MFCD22547077 CAS Number: 1393572-94-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...