2-Amino-5-(3-(piperazin-1-yl)phenyl)-N-(pyridin-4-yl)nicotinamide 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
PKC-iota inhibitor 1
A1363185-10MG
A1363185-1MG
A1363185-50MG
A1363185-5MG
76682-372EA
74.27
USD
76682-372
76682-374
76682-376
76682-378
2-Amino-5-(3-(piperazin-1-yl)phenyl)-N-(pyridin-4-yl)nicotinamide 98+%
2-Amino-5-(3-(piperazin-1-yl)phenyl)-N-(pyridin-4-yl)nicotinamide
Precaución: for Research Use Only
Formula:
C₂₁H₂₂N₆O MW: 374.4390 g/mol |
MDL Number:
MFCD32062834 CAS Number: 2328094-11-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...