[Ir(dFOMeppy)2-(5,5'-dCF3bpy)]PF6 97%

Supplier: Ambeed
Warning

A1362925-100MG A1362925-1G A1362925-250MG
76682-362EA 95 USD
76682-362 76682-364 76682-366
[Ir(dFOMeppy)2-(5,5'-dCF3bpy)]PF6 97%
[Ir(dFOMeppy)2-(5,5'-dCF3bpy)]PF6

Precaución: for Research Use Only

Formula: C₃₆H₂₂F₁₆IrN₄O₂P
MDL Number: MFCD32672288
CAS Number: 2307271-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR