[(1R,5S,6R)-3-Benzyl-3-azabicyclo[3.1.0]hexan-6-yl]methanol 97%

Supplier: Ambeed
Warning

Synonyms: (1R,5S,6R)-3-Aza-3-benzyl-6-(hydroxymethyl)bicyclo[3.1.0]hexane, rel-((1R,5S,6r)-3-Benzyl-3-azabicyclo[3.1.0]hexan-6-yl)methanol

A1362883-1G A1362883-25G A1362883-5G
76682-356EA 48.81 USD
76682-356 76682-358 76682-360
[(1R,5S,6R)-3-Benzyl-3-azabicyclo[3.1.0]hexan-6-yl]methanol 97%
[(1R,5S,6R)-3-Benzyl-3-azabicyclo[3.1.0]hexan-6-yl]methanol

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO
MW: 203.29 g/mol
MDL Number: MFCD16140332
CAS Number: 134575-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR