5-Bromo-7H-benzo[c]fluoren-7-one 97%

Supplier: Ambeed
Warning

A1362583-1G A1362583-250MG A1362583-5G
76682-350EA 18 USD
76682-350 76682-352 76682-354
5-Bromo-7H-benzo[c]fluoren-7-one 97%
5-Bromo-7H-benzo[c]fluoren-7-one

Precaución: for Research Use Only

Formula: C₁₇H₉BrO
CAS Number: 64356-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR