Benzyl ((6-chloro-3H-imidazo[4,5-b]pyridin-2-yl)methyl)carbamate 97%

Supplier: Ambeed
Warning

A1360445-100MG A1360445-250MG
76682-338EA 51 USD
76682-338 76682-340
Benzyl ((6-chloro-3H-imidazo[4,5-b]pyridin-2-yl)methyl)carbamate 97%
Benzyl ((6-chloro-3H-imidazo[4,5-b]pyridin-2-yl)methyl)carbamate

Precaución: for Research Use Only

Formula: C₁₅H₁₃ClN₄O₂
MDL Number: MFCD33022419
CAS Number: 2682112-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR