5-Chloro-N-(4-chlorobenzyl)-2-(ethylsulfonyl)-N-(furan-2-ylmethyl)pyrimidine-4-carboxamide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
ZAP-180013
A1359461-10MG
A1359461-1MG
A1359461-5MG
76682-328EA
84.88
USD
76682-328
76682-330
76682-332
5-Chloro-N-(4-chlorobenzyl)-2-(ethylsulfonyl)-N-(furan-2-ylmethyl)pyrimidine-4-carboxamide 98%
5-Chloro-N-(4-chlorobenzyl)-2-(ethylsulfonyl)-N-(furan-2-ylmethyl)pyrimidine-4-carboxamide
Precaución: for Research Use Only
Formula:
C₁₉H₁₇Cl₂N₃O₄S MW: 454.33 g/mol |
MDL Number:
MFCD07360519 CAS Number: 873080-25-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...