2-(4-Chloro-2-fluorophenyl)propan-2-ol 97%

Supplier: Ambeed
Warning

A1355156-1G A1355156-250MG
76682-300EA 211.12 USD
76682-300 76682-302
2-(4-Chloro-2-fluorophenyl)propan-2-ol 97%
2-(4-Chloro-2-fluorophenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₉H₁₀ClFO
MW: 188.6265 g/mol
MDL Number: MFCD23852786
CAS Number: 1537457-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR