Bis(triphenylphosphoranylidene)ammonium chloride 98%

Supplier: Ambeed
Warning

Synonyms: 1,1,1-Triphenyl-N-(triphenylphosphoranylidene)phosphoraniminium chloride, Bis(triphenylphosphine)iminium chloride

A1353874-25G A1353874-5G A1353874-100G
76682-296EA 54.11 USD
76682-296 76682-298 77596-852
Bis(triphenylphosphoranylidene)ammonium chloride 98%
Bis(triphenylphosphoranylidene)ammonium chloride

Precaución: for Research Use Only

Formula: C₃₆H₃₀ClNP₂
MW: 574.041 g/mol
MDL Number: MFCD00151523
CAS Number: 21050-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR