2-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-benzo[d][1,2,3]triazole 98%

Supplier: Ambeed
Warning

A1353798-1G A1353798-250MG
76682-292EA 606.84 USD
76682-292 76682-294
2-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-benzo[d][1,2,3]triazole 98%
2-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-benzo[d][1,2,3]triazole

Precaución: for Research Use Only

Formula: C₁₃H₁₈BN₃O₂
MW: 259.1119 g/mol
MDL Number: MFCD32661390
CAS Number: 2259877-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR