2,6-Bis((S)-4-isobutyl-4,5-dihydrooxazol-2-yl)pyridine ≥98%, ee 99%
Supplier: Ambeed
|
Warning
|
A1353133-100MG
A1353133-250MG
76682-284EA
104
USD
76682-284
76682-286
2,6-Bis((S)-4-isobutyl-4,5-dihydrooxazol-2-yl)pyridine ≥98%, ee 99%
2,6-Bis((S)-4-isobutyl-4,5-dihydrooxazol-2-yl)pyridine
Precaución: for Research Use Only
Formula:
C₁₉H₂₇N₃O₂ |
CAS Number:
335357-38-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...