(4R,4'R)-2,2'-(1,3-Diphenylpropane-2,2-diyl)bis(4-benzyl-4,5-dihydrooxazole) 98%

Supplier: Ambeed
Warning

A1353052-100MG A1353052-1G A1353052-250MG
76682-256EA 122.01 USD
76682-256 76682-258 76682-260
(4R,4'R)-2,2'-(1,3-Diphenylpropane-2,2-diyl)bis(4-benzyl-4,5-dihydrooxazole) 98%
(4R,4'R)-2,2'-(1,3-Diphenylpropane-2,2-diyl)bis(4-benzyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₃₅H₃₄N₂O₂
MW: 514.6567 g/mol
CAS Number: 2279976-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR