(4S,4'S)-4,4'-Dicyclohexyl-4,4',5,5'-tetrahydro-2,2'-bioxazole 98%

Supplier: Ambeed
Warning

A1353041-100MG A1353041-1G A1353041-250MG A1353041-5G
76682-250EA 37 USD
76682-250 76682-252 76682-254 77867-465
(4S,4'S)-4,4'-Dicyclohexyl-4,4',5,5'-tetrahydro-2,2'-bioxazole 98%
(4S,4'S)-4,4'-Dicyclohexyl-4,4',5,5'-tetrahydro-2,2'-bioxazole

Precaución: for Research Use Only

Formula: C₁₈H₂₈N₂O₂
MW: 304.4271 g/mol
CAS Number: 2368835-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR