Methyl 1,4-Dioxaspiro[4.5]decane-8-carboxylate 96%

Supplier: Ambeed
Warning

A1352869-100G A1352869-1G A1352869-5G A1352869-25G
76682-244EA 650 USD
76682-244 76682-246 76682-248 77867-463
Methyl 1,4-Dioxaspiro[4.5]decane-8-carboxylate 96%
Methyl 1,4-Dioxaspiro[4.5]decane-8-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₄
MDL Number: MFCD13195278
CAS Number: 26845-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR