(9H-Fluoren-9-yl)methyl (2-aminoethyl)carbamate hydrobromide 97%

Supplier: Ambeed
Warning

A1352827-100MG A1352827-250MG A1352827-1G
76682-238EA 94 USD
76682-238 76682-240 77867-462
(9H-Fluoren-9-yl)methyl (2-aminoethyl)carbamate hydrobromide 97%
(9H-Fluoren-9-yl)methyl (2-aminoethyl)carbamate hydrobromide

Precaución: for Research Use Only

Formula: C₁₇H₁₉BrN₂O₂
MW: 363.2490 g/mol
MDL Number: MFCD01863041
CAS Number: 352351-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR