1-(3-Fluorophenyl)-3-(3-((7-(1-methyl-1H-pyrazol-4-yl)quinazolin-4-yl)amino)phenyl)urea 97%

Supplier: Ambeed
Warning

Synonyms: SP-96

A1352783-10MG A1352783-1MG A1352783-25MG A1352783-50MG A1352783-5MG
76682-228EA 66.84 USD
76682-228 76682-230 76682-232 76682-234 76682-236
1-(3-Fluorophenyl)-3-(3-((7-(1-methyl-1H-pyrazol-4-yl)quinazolin-4-yl)amino)phenyl)urea 97%
1-(3-Fluorophenyl)-3-(3-((7-(1-methyl-1H-pyrazol-4-yl)quinazolin-4-yl)amino)phenyl)urea

Precaución: for Research Use Only

Formula: C₂₅H₂₀FN₇O
MW: 453.47 g/mol
MDL Number: MFCD32858307
CAS Number: 2682114-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR