Hexahydropyridazine dihydrochloride 95%

Supplier: Ambeed
Warning

A1352316-100G A1352316-1G A1352316-25G A1352316-5G
76682-218EA 303 USD
76682-218 76682-220 76682-222 76682-224
Hexahydropyridazine dihydrochloride 95%
Hexahydropyridazine dihydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₂Cl₂N₂
MW: 159.05 g/mol
MDL Number: MFCD09037903
CAS Number: 124072-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR