tert-Butyl 4-(allyloxy)piperidine-1-carboxylate 98% + stabilized
Supplier: Ambeed
|
Warning
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
A1352105-1G
A1352105-250MG
A1352105-5G
76682-212EA
75.99
USD
76682-212
76682-214
76682-216
tert-Butyl 4-(allyloxy)piperidine-1-carboxylate 98% + stabilized
tert-Butyl 4-(allyloxy)piperidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₃H₂₃NO₃ |
MDL Number:
MFCD09261170 CAS Number: 163210-43-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...