(S)-2-([2,2'-Bipyridin]-6-yl)-4-benzyl-4,5-dihydrooxazole ≥97%, ee 99%

Supplier: Ambeed
Warning

A1351975-100MG A1351975-1G A1351975-250MG
76682-194EA 78 USD
76682-194 76682-196 76682-198
(S)-2-([2,2'-Bipyridin]-6-yl)-4-benzyl-4,5-dihydrooxazole ≥97%, ee 99%
(S)-2-([2,2'-Bipyridin]-6-yl)-4-benzyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₀H₁₇N₃O
MW: 315.3685 g/mol
CAS Number: 2757082-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR