4,4'-(1,2-Diphenylethene-1,2-diyl)dibenzonitrile 97%

Supplier: Ambeed
Warning

A1351853-1G A1351853-250MG A1351853-5G
76682-146EA 34 USD
76682-146 76682-148 76682-150
4,4'-(1,2-Diphenylethene-1,2-diyl)dibenzonitrile 97%
4,4'-(1,2-Diphenylethene-1,2-diyl)dibenzonitrile

Precaución: for Research Use Only

Formula: C₂₈H₁₈N₂
CAS Number: 2244891-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR