4,4'-(2,2-Bis(4-bromophenyl)ethene-1,1-diyl)diphenol 97%

Supplier: Ambeed
Warning

A1351851-100MG A1351851-1G A1351851-250MG
76682-140EA 51.99 USD
76682-140 76682-142 76682-144
4,4'-(2,2-Bis(4-bromophenyl)ethene-1,1-diyl)diphenol 97%
4,4'-(2,2-Bis(4-bromophenyl)ethene-1,1-diyl)diphenol

Precaución: for Research Use Only

Formula: C₂₆H₁₈Br₂O₂
MW: 522.2279 g/mol
CAS Number: 2029185-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR