(4-(1,2,2-Triphenylvinyl)phenyl)methanol 97%

Supplier: Ambeed
Warning

A1351846-1G A1351846-250MG A1351846-5G
76682-134EA 95.49 USD
76682-134 76682-136 76682-138
(4-(1,2,2-Triphenylvinyl)phenyl)methanol 97%
(4-(1,2,2-Triphenylvinyl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₂₇H₂₂O
MW: 362.4630 g/mol
MDL Number: MFCD30470533
CAS Number: 1015082-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR