(2S,2'S)-2,2'-(1,3,6,8-Tetraoxo-1,3,6,8-tetrahydrobenzo[lmn][3,8]phenanthroline-2,7-diyl)dipropionic acid 97%

Supplier: Ambeed
Warning

A1351823-1G A1351823-250MG A1351823-5G
76682-128EA 53.05 USD
76682-128 76682-130 76682-132
(2S,2'S)-2,2'-(1,3,6,8-Tetraoxo-1,3,6,8-tetrahydrobenzo[lmn][3,8]phenanthroline-2,7-diyl)dipropionic acid 97%
(2S,2'S)-2,2'-(1,3,6,8-Tetraoxo-1,3,6,8-tetrahydrobenzo[lmn][3,8]phenanthroline-2,7-diyl)dipropionic acid

Precaución: for Research Use Only

Formula: C₂₀H₁₄N₂O₈
MW: 410.3338 g/mol
CAS Number: 429692-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR