4,4''-Dihydroxy-[1,1':4',1''-terphenyl]-3,3''-dicarboxylic acid 97%

Supplier: Ambeed
Warning

A1351815-1G A1351815-5G
76682-116EA 254.62 USD
76682-116 76682-118
4,4''-Dihydroxy-[1,1':4',1''-terphenyl]-3,3''-dicarboxylic acid 97%
4,4''-Dihydroxy-[1,1':4',1''-terphenyl]-3,3''-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₂₀H₁₄O₆
MW: 350.3216 g/mol
CAS Number: 1623061-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR