5,5-Difluoro-2,8-diiodo-1,3,7,9,10-pentamethyl-5H-4l4,5l4-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinine 97%

Supplier: Ambeed
Warning

Synonyms: 2,6-Diiodo-BODIPY 493/503

A1351809-100MG A1351809-1G A1351809-250MG
76682-114EA 71 USD
76682-114 77867-433 77867-434
5,5-Difluoro-2,8-diiodo-1,3,7,9,10-pentamethyl-5H-4l4,5l4-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinine 97%
5,5-Difluoro-2,8-diiodo-1,3,7,9,10-pentamethyl-5H-4l4,5l4-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinine

Precaución: for Research Use Only

Formula: C₁₄H₁₅BF₂I₂N₂
MW: 513.8991 g/mol
CAS Number: 1031443-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR