2'-(tert-Butyl(phenyl)phosphino)-N2,N2,N6,N6-tetramethyl-[1,1'-biphenyl]-2,6-diamine 98%

Supplier: Ambeed
Warning

A1351253-100MG A1351253-1G A1351253-250MG A1351253-5G
76682-090EA 40 USD
76682-090 76682-092 76682-094 77621-634
2'-(tert-Butyl(phenyl)phosphino)-N2,N2,N6,N6-tetramethyl-[1,1'-biphenyl]-2,6-diamine 98%
2'-(tert-Butyl(phenyl)phosphino)-N2,N2,N6,N6-tetramethyl-[1,1'-biphenyl]-2,6-diamine

Precaución: for Research Use Only

Formula: C₂₆H₃₃N₂P
MDL Number: MFCD29905429
CAS Number: 1660153-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR