2-(3-Amino-3-oxopropyl)malonic acid 97%

Supplier: Ambeed
Warning

A1351243-100MG A1351243-1G A1351243-250MG
76682-084EA 60.48 USD
76682-084 76682-086 76682-088
2-(3-Amino-3-oxopropyl)malonic acid 97%
2-(3-Amino-3-oxopropyl)malonic acid

Precaución: for Research Use Only

Formula: C₆H₉NO₅
MW: 175.1394 g/mol
MDL Number: MFCD33022391
CAS Number: 2682114-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR