Dichlorobis(dimethylglyoxime)cobalt(II) 97%

Supplier: Ambeed
Danger

A1350122-1G A1350122-250MG A1350122-5G
76682-074EA 53 USD
76682-074 76682-076 76682-078
Dichlorobis(dimethylglyoxime)cobalt(II) 97%
Dichlorobis(dimethylglyoxime)cobalt(II)

Precaución: for Research Use Only

Formula: C₈H₁₆Cl₂CoN₄O₄
MW: 362.0762 g/mol
CAS Number: 14784-26-0
UN: 1325
ADR: 4.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR