Ethyl-3-oxo-3-(pyridin-4-yl)propanoate 98%

Supplier: Ambeed
Warning

A134763-10G A134763-1G A134763-250MG A134763-25G A134763-5G
76682-050EA 29 USD
76682-050 76682-052 76682-054 76682-056 76682-058
Ethyl-3-oxo-3-(pyridin-4-yl)propanoate 98%
Ethyl-3-oxo-3-(pyridin-4-yl)propanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₃
MDL Number: MFCD00094021
CAS Number: 26377-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR