2-Benzyl-1H-indole 98%

Supplier: Ambeed
Warning

A1340715-100MG A1340715-1G A1340715-250MG
76682-024EA 755 USD
76682-024 76682-026 76682-028
2-Benzyl-1H-indole 98%
2-Benzyl-1H-indole

Precaución: for Research Use Only

Formula: C₁₅H₁₃N
MDL Number: MFCD18449615
CAS Number: 3377-72-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR