3,5-Bis(trifluoromethyl)pyridin-2(1H)-one 97%

Supplier: Ambeed
Warning

A1339666-100MG A1339666-250MG
76682-014EA 101.85 USD
76682-014 77621-594
3,5-Bis(trifluoromethyl)pyridin-2(1H)-one 97%
3,5-Bis(trifluoromethyl)pyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₇H₃F₆NO
MW: 231.1 g/mol
MDL Number: MFCD28802532
CAS Number: 38609-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR