2-(8-(Benzyloxy)imidazo[1,2-a]pyridin-2-yl)acetic acid hydrochloride 97%

Supplier: Ambeed
Warning

A1339450-100MG A1339450-1G A1339450-250MG
76681-994EA 58.35 USD
76681-994 76681-996 76681-998
2-(8-(Benzyloxy)imidazo[1,2-a]pyridin-2-yl)acetic acid hydrochloride 97%
2-(8-(Benzyloxy)imidazo[1,2-a]pyridin-2-yl)acetic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₆H₁₅ClN₂O₃
MW: 318.7549 g/mol
MDL Number: MFCD32856790
CAS Number: 2682112-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR