(4-Iodophenyl)diphenylphosphane 98%

Supplier: Ambeed
Warning

A1339360-100MG A1339360-250MG
76681-990EA 37.14 USD
76681-990 76681-992
(4-Iodophenyl)diphenylphosphane 98%
(4-Iodophenyl)diphenylphosphane

Precaución: for Research Use Only

Formula: C₁₈H₁₄IP
MW: 388.1820 g/mol
CAS Number: 440111-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR