2-Butyl-5,5-dimethyl-1,3,2-dioxaborinane 98%
Supplier: Ambeed
|
Warning
|
A1338817-1G
A1338817-5G
76681-976EA
18.04
USD
76681-976
76681-978
2-Butyl-5,5-dimethyl-1,3,2-dioxaborinane 98%
2-Butyl-5,5-dimethyl-1,3,2-dioxaborinane
Precaución: for Research Use Only
Formula:
C₉H₁₉BO₂ MW: 170.0570 g/mol |
CAS Number:
879224-89-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...