(2-(4-Fluorophenyl)cyclopropyl)methanamine hydrochloride 98%

Supplier: Ambeed
Warning

A1337454-100MG A1337454-1G A1337454-250MG
76681-970EA 94 USD
76681-970 76681-972 76681-974
(2-(4-Fluorophenyl)cyclopropyl)methanamine hydrochloride 98%
(2-(4-Fluorophenyl)cyclopropyl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₃ClFN
MDL Number: MFCD20502100
CAS Number: 1354954-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR