m-Toluenesulfonic acid monohydrate ≥98%

Supplier: Ambeed
Danger

A1329955-1G A1329955-250MG
76681-962EA 78 USD
76681-962 76681-964
m-Toluenesulfonic acid monohydrate ≥98%
m-Toluenesulfonic acid monohydrate

Precaución: for Research Use Only

Formula: C₇H₈O₃S·H₂O
MDL Number: MFCD00167130
CAS Number: 312619-56-0
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR