Tetra-n-butylammonium Di-tert-butylphosphate 95% (Ammonium:phosphate ~1.2-1.5)

Supplier: Ambeed
Warning

A1329937-100G A1329937-1G A1329937-250MG A1329937-25G A1329937-5G
76681-952EA 271 USD
76681-952 76681-954 76681-956 76681-958 76681-960
Tetra-n-butylammonium Di-tert-butylphosphate 95% (Ammonium:phosphate ~1.2-1.5)
Tetra-n-butylammonium Di-tert-butylphosphate

Precaución: for Research Use Only

Formula: C₂₄H₅₄NO₄P
MDL Number: MFCD12198885
CAS Number: 68695-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR