Ailver(I) pivalate 97%

Supplier: Ambeed
Warning

A1329714-250MG A1329714-1G A1329714-100MG A1329714-50MG
76681-950EA 211.12 USD
76681-950 77566-598 77621-568 77621-570
Ailver(I) pivalate 97%
Ailver(I) pivalate

Precaución: For research use only.

Formula: C₅H₉AgO₂
MW: 208.99 g/mol
MDL Number: MFCD34187019
CAS Number: 7324-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR