2-(2,4-Dichlorophenyl)-2-fluoroethanamine hydrochloride 97%

Supplier: Ambeed
Warning

A1329704-100MG A1329704-1G A1329704-250MG
76681-944EA 82.76 USD
76681-944 76681-946 76681-948
2-(2,4-Dichlorophenyl)-2-fluoroethanamine hydrochloride 97%
2-(2,4-Dichlorophenyl)-2-fluoroethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₉Cl₃FN
MW: 244.5212 g/mol
CAS Number: 960053-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR