2-Amino-3-(8-hydroxyquinolin-5-yl)propanoic acid dihydrochloride 97%

Supplier: Ambeed
Warning

A1329490-100MG A1329490-250MG A1329490-1G
76681-938EA 176 USD
76681-938 76681-940 77867-341
2-Amino-3-(8-hydroxyquinolin-5-yl)propanoic acid dihydrochloride 97%
2-Amino-3-(8-hydroxyquinolin-5-yl)propanoic acid dihydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₄Cl₂N₂O₃
MW: 305.1572 g/mol
CAS Number: 23218-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR