3-(3-Methyl-1H-pyrazol-5-yl)pyridine 97%

Supplier: Ambeed
Warning

A1329285-100MG A1329285-1G A1329285-250MG
76681-932EA 42 USD
76681-932 76681-934 76681-936
3-(3-Methyl-1H-pyrazol-5-yl)pyridine 97%
3-(3-Methyl-1H-pyrazol-5-yl)pyridine
Formula: C₉H₉N₃
MDL Number: MFCD01436332
CAS Number: 19959-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR