Ir[FCF3(CF3)ppy]2(dtbbpy)PF6 98%

Supplier: Ambeed
Warning

A1329209-100MG A1329209-1G A1329209-250MG
76681-914EA 148.53 USD
76681-914 76681-916 76681-918
Ir[FCF3(CF3)ppy]2(dtbbpy)PF6 98%
Ir[FCF3(CF3)ppy]2(dtbbpy)PF6

Precaución: for Research Use Only

Formula: C₄₄H₃₄F₂₀IrN₄P
MW: 1221.9264 g/mol
CAS Number: 2222402-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR